Assay Details
| Gene Symbol: ALiX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143325.4 | NP_651582.1 | 4 - 5 | 2337 | 73 | |
| GenBank mRNA | AY069534.1 | - | 4 - 5 | 2435 | 73 | |
More Information
Set Membership: |
|

ALiX
Genomic Map