Assay Details
| Gene Symbol: mino | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260414.2 | NP_001247343.2 | 2 - 3 | 448 | 62 | |
| NM_143340.4 | NP_651597.1 | 2 - 3 | 922 | 62 | ||
| GenBank mRNA | AF145635.1 | - | 2 - 3 | 910 | 62 | |
More Information
Set Membership: |
|

mino
Genomic Map