Assay Details
| Gene Symbol: mino | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260414.2 | NP_001247343.2 | 4 - 5 | 1602 | 67 | |
| NM_143340.4 | NP_651597.1 | 4 - 5 | 2076 | 67 | ||
| NM_170358.2 | NP_733237.1 | 3 - 4 | 1433 | 67 | ||
| NM_170359.3 | NP_733238.1 | 3 - 4 | 1322 | 67 | ||
| GenBank mRNA | AF145635.1 | - | 4 - 5 | 2064 | 67 | |
| BT124983.1 | - | 5 - 6 | 2292 | 67 | ||
More Information
Set Membership: |
|

mino
Genomic Map