Assay Details
| Gene Symbol: BOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170280.2 | NP_001163751.1 | 1 - 2 | 353 | 75 | |
| NM_001276101.1 | NP_001263030.1 | 1 - 2 | 353 | 75 | ||
| NM_001276102.1 | NP_001263031.1 | 1 - 2 | 353 | 75 | ||
| NM_143342.2 | NP_651599.1 | 2 - 3 | 608 | 75 | ||
| NM_170361.3 | NP_733240.1 | 1 - 2 | 353 | 75 | ||
| GenBank mRNA | BT032836.1 | - | 1 - 2 | 284 | 75 | |
More Information
Set Membership: |
|

BOD1
Genomic Map