Assay Details
| Gene Symbol: CG4849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143348.4 | NP_651605.1 | 3 - 4 | 935 | 106 | |
| GenBank mRNA | AY089551.1 | - | 3 - 4 | 913 | 106 | |
More Information
Set Membership: |
|

CG4849
Genomic Map