Assay Details
| Gene Symbol: CG4849 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143348.4 | NP_651605.1 | 6 - 7 | 2721 | 112 | |
| GenBank mRNA | AY089551.1 | - | 6 - 7 | 2699 | 112 | |
More Information
Set Membership: |
|

CG4849
Genomic Map