Assay Details
| Gene Symbol: trv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170283.3 | NP_001163754.3 | 5 - 6 | 1539 | 82 | |
| NM_001276103.1 | NP_001263032.1 | 4 - 5 | 1305 | 82 | ||
| NM_001276104.1 | NP_001263033.1 | 4 - 5 | 1305 | 82 | ||
| NM_001300644.1 | NP_001287573.1 | 4 - 5 | 1305 | 82 | ||
| NM_143352.4 | NP_651609.1 | 4 - 5 | 1305 | 82 | ||
| GenBank mRNA | AY121663.1 | - | 4 - 5 | 1303 | 82 | |
More Information
Set Membership: |
|

trv
Genomic Map