Assay Details
| Gene Symbol: trv | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170283.3 | NP_001163754.3 | 9 - 10 | 2374 | 62 | |
| NM_143352.4 | NP_651609.1 | 8 - 9 | 2140 | 62 | ||
| GenBank mRNA | AY121663.1 | - | 8 - 9 | 2138 | 62 | |
More Information
Set Membership: |
|

trv
Genomic Map