Assay Details
| Gene Symbol: UQCR-14L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143357.3 | NP_651614.1 | 1 | 120 | 59 | |
| GenBank mRNA | KX531464.1 | - | 1 | 34 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UQCR-14L| Gene Symbol: UQCR-14L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143357.3 | NP_651614.1 | 1 | 120 | 59 | |
| GenBank mRNA | KX531464.1 | - | 1 | 34 | 59 | |
Set Membership: |
|