Assay Details
| Gene Symbol: mIF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143364.3 | NP_651621.2 | 3 - 4 | 923 | 78 | |
| GenBank mRNA | BT150258.1 | - | 3 - 4 | 900 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mIF2| Gene Symbol: mIF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143364.3 | NP_651621.2 | 3 - 4 | 923 | 78 | |
| GenBank mRNA | BT150258.1 | - | 3 - 4 | 900 | 78 | |
Set Membership: |
|