Assay Details
| Gene Symbol: rdog | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143421.3 | NP_651678.1 | 2 - 3 | 799 | 57 | |
| NM_170399.2 | NP_733278.1 | 3 - 4 | 648 | 57 | ||
| GenBank mRNA | AY051660.1 | - | 2 - 3 | 760 | 57 | |
More Information
Set Membership: |
|

rdog
Genomic Map