Assay Details
| Gene Symbol: alph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143444.4 | NP_651701.1 | 6 - 7 | 1577 | 67 | |
| GenBank mRNA | AY051685.1 | - | 6 - 7 | 1553 | 67 | |
More Information
Set Membership: |
|

alph
Genomic Map