Assay Details
| Gene Symbol: Kul | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300660.1 | NP_001287589.1 | 7 - 8 | 2993 | 65 | |
| NM_001300661.1 | NP_001287590.1 | 7 - 8 | 2947 | 65 | ||
| NM_143459.3 | NP_651716.1 | 7 - 8 | 2947 | 65 | ||
More Information
Set Membership: |
|

Kul
Genomic Map