Assay Details
| Gene Symbol: Vps16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143474.3 | NP_651731.1 | 1 | 1348 | 79 | |
| GenBank mRNA | BT003209.1 | - | 1 | 1346 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vps16B| Gene Symbol: Vps16B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143474.3 | NP_651731.1 | 1 | 1348 | 79 | |
| GenBank mRNA | BT003209.1 | - | 1 | 1346 | 79 | |
Set Membership: |
|