Assay Details
| Gene Symbol: RpS8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260433.3 | NP_001247362.1 | 2 - 3 | 174 | 80 | |
| NM_001300667.1 | NP_001287596.1 | 2 - 3 | 144 | 80 | ||
| NM_001300668.1 | NP_001287597.1 | 3 - 4 | 349 | 80 | ||
| NM_143483.3 | NP_651740.1 | 2 - 3 | 144 | 80 | ||
| GenBank mRNA | AY118424.1 | - | 2 - 3 | 174 | 80 | |
| AY118446.1 | - | 1995 | 80 | |||
More Information
Set Membership: |
|

RpS8
Genomic Map