Assay Details
| Gene Symbol: eIF2Balpha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143495.3 | NP_651752.1 | 3 - 4 | 414 | 72 | |
| GenBank mRNA | AJ344146.1 | - | 3 - 4 | 388 | 72 | |
| AY060991.1 | - | 3 - 4 | 388 | 72 | ||
More Information
Set Membership: |
|

eIF2Balpha
Genomic Map