Assay Details
| Gene Symbol: CG9737 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143526.2 | NP_651783.1 | 1 - 2 | 631 | 81 | |
| GenBank mRNA | AY119642.1 | - | 1 - 2 | 629 | 81 | |
More Information
Set Membership: |
|

CG9737
Genomic Map