Assay Details
| Gene Symbol: aralar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276178.1 | NP_001263107.1 | 1 - 2 | 185 | 76 | |
| NM_143538.3 | NP_651795.2 | 1 - 2 | 185 | 76 | ||
| NM_170486.2 | NP_733365.1 | 1 - 2 | 185 | 76 | ||
| GenBank mRNA | AY058654.1 | - | 1 - 2 | 150 | 76 | |
More Information
Set Membership: |
|

aralar1
Genomic Map