Assay Details
| Gene Symbol: aralar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260439.1 | NP_001247368.1 | 3 - 4 | 528 | 115 | |
| NM_001276178.1 | NP_001263107.1 | 3 - 4 | 489 | 115 | ||
| NM_143538.3 | NP_651795.2 | 3 - 4 | 489 | 115 | ||
| NM_170485.2 | NP_733364.1 | 3 - 4 | 528 | 115 | ||
| NM_170486.2 | NP_733365.1 | 3 - 4 | 489 | 115 | ||
| NM_170487.3 | NP_733366.2 | 3 - 4 | 664 | 115 | ||
| GenBank mRNA | AY058654.1 | - | 3 - 4 | 454 | 115 | |
More Information
Set Membership: |
|

aralar1
Genomic Map