Assay Details
| Gene Symbol: aralar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260439.1 | NP_001247368.1 | 8 - 9 | 1242 | 97 | |
| NM_001276178.1 | NP_001263107.1 | 7 - 8 | 1167 | 97 | ||
| NM_143538.3 | NP_651795.2 | 7 - 8 | 1167 | 97 | ||
| NM_170485.2 | NP_733364.1 | 7 - 8 | 1206 | 97 | ||
| NM_170486.2 | NP_733365.1 | 7 - 8 | 1167 | 97 | ||
| NM_170487.3 | NP_733366.2 | 7 - 8 | 1342 | 97 | ||
| GenBank mRNA | AY058654.1 | - | 7 - 8 | 1132 | 97 | |
More Information
Set Membership: |
|

aralar1
Genomic Map