Assay Details
| Gene Symbol: cindr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276195.1 | NP_001263124.1 | 5 - 6 | 2288 | 91 | |
| NM_001276196.1 | NP_001263125.1 | 3 - 4 | 1370 | 91 | ||
| NM_001276197.1 | NP_001263126.1 | 3 - 4 | 1376 | 91 | ||
| NM_001276198.1 | NP_001263127.1 | 2 - 3 | 681 | 91 | ||
| NM_001276199.1 | NP_001263128.1 | 2 - 3 | 625 | 91 | ||
| NM_001276200.1 | NP_001263129.1 | 6 - 7 | 2385 | 91 | ||
| NM_143570.4 | NP_651827.1 | 2 - 3 | 707 | 91 | ||
| NM_170526.3 | NP_733405.2 | 5 - 6 | 2208 | 91 | ||
| NM_170527.3 | NP_733406.1 | 2 - 3 | 707 | 91 | ||
| NM_170528.3 | NP_733407.2 | 2 - 3 | 1199 | 91 | ||
| GenBank mRNA | AF145614.1 | - | 2 - 3 | 685 | 91 | |
More Information
Set Membership: |
|

cindr
Genomic Map