Assay Details
| Gene Symbol: pasha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276221.1 | NP_001263150.1 | 1 - 2 | 349 | 71 | |
| NM_143622.3 | NP_651879.1 | 1 - 2 | 800 | 71 | ||
| GenBank mRNA | AY058524.1 | - | 1 - 2 | 794 | 71 | |
More Information
Set Membership: |
|

pasha
Genomic Map