Assay Details
| Gene Symbol: pasha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276220.1 | NP_001263149.1 | 3 - 4 | 1222 | 64 | |
| NM_001276221.1 | NP_001263150.1 | 3 - 4 | 1495 | 64 | ||
| NM_143622.3 | NP_651879.1 | 3 - 4 | 1946 | 64 | ||
| GenBank mRNA | AY058524.1 | - | 3 - 4 | 1940 | 64 | |
More Information
Set Membership: |
|

pasha
Genomic Map