Assay Details
| Gene Symbol: nero | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143630.2 | NP_651887.1 | 1 | 1058 | 93 | |
| GenBank mRNA | AY118500.1 | - | 1 | 1016 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

nero| Gene Symbol: nero | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143630.2 | NP_651887.1 | 1 | 1058 | 93 | |
| GenBank mRNA | AY118500.1 | - | 1 | 1016 | 93 | |
Set Membership: |
|