Assay Details
Gene Symbol: CG2187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202402.1 | NP_001189331.1 | 7 - 8 | 1582 | 72 | |
NM_143633.2 | NP_651890.1 | 7 - 8 | 2141 | 72 | ||
GenBank mRNA | U72716.1 | - | 7 - 8 | 2142 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
