Assay Details
| Gene Symbol: betaGlu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143636.3 | NP_651893.1 | 2 - 3 | 1575 | 69 | |
| GenBank mRNA | BT010073.1 | - | 2 - 3 | 1548 | 69 | |
More Information
Set Membership: |
|

betaGlu
Genomic Map