Assay Details
| Gene Symbol: sip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276223.2 | NP_001263152.1 | 4 - 5 | 1561 | 64 | |
| NM_143637.5 | NP_651894.3 | 4 - 5 | 1561 | 64 | ||
| GenBank mRNA | AY060677.1 | - | 4 - 5 | 1542 | 64 | |
More Information
Set Membership: |
|

sip3
Genomic Map