Assay Details
| Gene Symbol: RhoGAP100F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170314.2 | NP_001163785.1 | 3 - 4 | 1356 | 68 | |
| NM_001170315.3 | NP_001163786.1 | 3 - 4 | 2108 | 68 | ||
| NM_001260465.1 | NP_001247394.1 | 3 - 4 | 1433 | 68 | ||
| NM_001260466.1 | NP_001247395.1 | 3 - 4 | 1433 | 68 | ||
| NM_001260467.2 | NP_001247396.1 | 3 - 4 | 1433 | 68 | ||
| NM_001260468.2 | NP_001247397.2 | 3 - 4 | 2108 | 68 | ||
| NM_001300700.1 | NP_001287629.1 | 3 - 4 | 2108 | 68 | ||
| NM_143645.2 | NP_651902.2 | 2 - 3 | 691 | 68 | ||
| GenBank mRNA | BT133435.1 | - | 3 - 4 | 1356 | 68 | |
More Information
Set Membership: |
|

RhoGAP100F
Genomic Map