Assay Details
| Gene Symbol: MED6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170106.1 | NP_001163577.1 | 3 - 4 | 971 | 62 | |
| NM_001170107.1 | NP_001163578.1 | 3 - 4 | 1015 | 62 | ||
| NM_001300335.1 | NP_001287264.1 | 2 - 3 | 818 | 62 | ||
| NM_143731.6 | NP_651988.2 | 3 - 4 | 1020 | 62 | ||
| NM_169310.2 | NP_731403.2 | 3 - 4 | 959 | 62 | ||
| GenBank mRNA | AY118517.1 | - | 3 - 4 | 997 | 62 | |
| BT001633.1 | - | 3 - 4 | 959 | 62 | ||
More Information
Set Membership: |
|

MED6
Genomic Map