Assay Details
| Gene Symbol: Invadolysin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143815.4 | NP_652072.1 | 1 - 2 | 865 | 68 | |
| GenBank mRNA | AF234630.1 | - | 1 - 2 | 132 | 68 | |
| BT001861.1 | - | 1 - 2 | 834 | 68 | ||
More Information
Set Membership: |
|

Invadolysin
Genomic Map