Assay Details
| Gene Symbol: Invadolysin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143815.4 | NP_652072.1 | 5 - 6 | 1236 | 67 | |
| GenBank mRNA | AF234630.1 | - | 5 - 6 | 503 | 67 | |
| BT001861.1 | - | 5 - 6 | 1205 | 67 | ||
More Information
Set Membership: |
|

Invadolysin
Genomic Map