Assay Details
| Gene Symbol: Invadolysin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143815.4 | NP_652072.1 | 8 - 9 | 2325 | 65 | |
| GenBank mRNA | AF234630.1 | - | 8 - 9 | 1592 | 65 | |
| BT001861.1 | - | 8 - 9 | 2294 | 65 | ||
More Information
Set Membership: |
|

Invadolysin
Genomic Map