Assay Details
| Gene Symbol: sqd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260159.2 | NP_001247088.1 | 6 - 7 | 1399 | 89 | |
| NM_143952.2 | NP_652209.1 | 6 - 7 | 1124 | 89 | ||
| NM_169528.2 | NP_731825.1 | 7 - 8 | 1507 | 89 | ||
| NM_206481.2 | NP_996203.1 | 8 - 9 | 1719 | 89 | ||
| GenBank mRNA | BT001384.1 | - | 8 - 9 | 1439 | 89 | |
| BT003283.1 | - | 8 - 9 | 1692 | 89 | ||
| BT023832.1 | - | 6 - 7 | 1107 | 89 | ||
More Information
Set Membership: |
|

sqd
Genomic Map