Assay Details
| Gene Symbol: pont | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144351.4 | NP_652608.1 | 1 - 2 | 1036 | 98 | |
| GenBank mRNA | AY061095.1 | - | 1 - 2 | 1047 | 98 | |
More Information
Set Membership: |
|

pont
Genomic Map