Assay Details
| Gene Symbol: Ugt303A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275541.1 | NP_001262470.1 | 1 - 2 | 300 | 86 | |
| NM_144364.2 | NP_652621.1 | 1 - 2 | 300 | 86 | ||
More Information
Set Membership: |
|

Ugt303A1
Genomic Map