Assay Details
| Gene Symbol: Ugt302C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144369.3 | NP_652626.1 | 2 - 3 | 662 | 68 | |
| GenBank mRNA | AY060891.1 | - | 2 - 3 | 638 | 68 | |
| BT006007.1 | - | 2 - 3 | 660 | 68 | ||
More Information
Set Membership: |
|

Ugt302C1
Genomic Map