Assay Details
| Gene Symbol: Nmdar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169059.2 | NP_730940.1 | 4 - 5 | 764 | 59 | |
| GenBank mRNA | AY070577.1 | - | 4 - 5 | 760 | 59 | |
| X71790.1 | - | 672 | 59 | |||
More Information
Set Membership: |
|

Nmdar1
Genomic Map