Assay Details
| Gene Symbol: Sec23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260003.1 | NP_001246932.1 | 3 - 4 | 1729 | 65 | |
| NM_001260004.1 | NP_001246933.1 | 3 - 4 | 1729 | 65 | ||
| NM_001260005.2 | NP_001246934.1 | 2 - 3 | 1850 | 65 | ||
| NM_001275365.1 | NP_001262294.1 | 3 - 4 | 1642 | 65 | ||
| NM_001300255.1 | NP_001287184.1 | 3 - 4 | 1729 | 65 | ||
| NM_169088.2 | NP_730978.1 | 3 - 4 | 1758 | 65 | ||
| NM_169089.4 | NP_730979.1 | 2 - 3 | 1850 | 65 | ||
More Information
Set Membership: |
|

Sec23
Genomic Map