Assay Details
| Gene Symbol: Pak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144541.2 | NP_001138013.2 | 4 - 5 | 1042 | 111 | |
| NM_001275380.2 | NP_001262309.1 | 4 - 5 | 744 | 111 | ||
| NM_001275381.2 | NP_001262310.1 | 4 - 5 | 749 | 111 | ||
| NM_001275382.1 | NP_001262311.1 | 4 - 5 | 1003 | 111 | ||
| NM_001275383.1 | NP_001262312.1 | 4 - 5 | 744 | 111 | ||
| NM_001275384.1 | NP_001262313.1 | 4 - 5 | 1042 | 111 | ||
| NM_079942.4 | NP_524681.1 | 4 - 5 | 749 | 111 | ||
| NM_169136.3 | NP_731073.1 | 4 - 5 | 1042 | 111 | ||
| NM_169137.3 | NP_731074.1 | 4 - 5 | 645 | 111 | ||
| GenBank mRNA | BT010291.1 | - | 4 - 5 | 1010 | 111 | |
More Information
Set Membership: |
|

Pak
Genomic Map