Assay Details
| Gene Symbol: CG42724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202255.1 | NP_001189184.1 | 6 - 7 | 3116 | 90 | |
| NM_001202257.3 | NP_001189186.1 | 8 - 9 | 3464 | 90 | ||
| NM_001275385.1 | NP_001262314.1 | 6 - 7 | 3017 | 90 | ||
| NM_001300264.1 | NP_001287193.1 | 7 - 8 | 3581 | 90 | ||
| NM_001300265.1 | NP_001287194.1 | 6 - 7 | 4673 | 90 | ||
More Information
Set Membership: |
|

CG42724
Genomic Map