Assay Details
| Gene Symbol: Alh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104227.3 | NP_001097697.2 | 1 - 2 | 1348 | 71 | |
| NM_001104228.3 | NP_001097698.1 | 1 - 2 | 1348 | 71 | ||
| NM_001170065.2 | NP_001163536.1 | 1 - 2 | 1348 | 71 | ||
| NM_001170066.2 | NP_001163537.1 | 1 - 2 | 1348 | 71 | ||
| NM_001275401.1 | NP_001262330.1 | 1 - 2 | 1348 | 71 | ||
| NM_001275402.1 | NP_001262331.1 | 1 - 2 | 1348 | 71 | ||
| NM_169170.3 | NP_731131.2 | 1 - 2 | 1348 | 71 | ||
| NM_169172.3 | NP_731133.1 | 1 - 2 | 1348 | 71 | ||
| GenBank mRNA | AY051905.1 | - | 1 - 2 | 1311 | 71 | |
More Information
Set Membership: |
|

Alh
Genomic Map