Assay Details
| Gene Symbol: Alh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275401.1 | NP_001262330.1 | 8 - 9 | 3911 | 85 | |
| NM_169170.3 | NP_731131.2 | 8 - 9 | 4007 | 85 | ||
More Information
Set Membership: |
|

Alh
Genomic Map