Assay Details
| Gene Symbol: DppIII | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169217.3 | NP_731237.1 | 3 - 4 | 604 | 56 | |
| NM_169218.3 | NP_731238.1 | 2 - 3 | 389 | 56 | ||
| GenBank mRNA | BT001283.1 | - | 3 - 4 | 539 | 56 | |
| BT001401.1 | - | 1 - 2 | 313 | 56 | ||
| BT001402.1 | - | 1 - 2 | 313 | 56 | ||
| BT001639.1 | - | 2 - 3 | 386 | 56 | ||
More Information
Set Membership: |
|

DppIII
Genomic Map