Assay Details
| Gene Symbol: mRpS18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169226.2 | NP_731252.1 | 1 - 2 | 189 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

mRpS18A| Gene Symbol: mRpS18A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169226.2 | NP_731252.1 | 1 - 2 | 189 | 69 | |
Set Membership: |
|