Assay Details
| Gene Symbol: pyd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260069.2 | NP_001246998.1 | 13 - 14 | 7283 | 87 | |
| NM_169245.3 | NP_731291.2 | 12 - 13 | 6375 | 87 | ||
More Information
Set Membership: |
|

pyd
Genomic Map