Assay Details
| Gene Symbol: ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043226.1 | NP_001036691.1 | 1 - 2 | 288 | 108 | |
| NM_169284.2 | NP_731355.1 | 1 - 2 | 288 | 108 | ||
More Information
Set Membership: |
|

ps
Genomic Map