Assay Details
| Gene Symbol: Syn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169332.4 | NP_731457.2 | 11 - 12 | 1734 | 103 | |
| NM_169333.3 | NP_731458.2 | 11 - 12 | 1734 | 103 | ||
| NM_169334.3 | NP_731459.2 | 11 - 12 | 1734 | 103 | ||
| NM_176451.3 | NP_788628.2 | 11 - 12 | 1734 | 103 | ||
More Information
Set Membership: |
|

Syn
Genomic Map