Assay Details
| Gene Symbol: hth | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014613.3 | NP_001014613.2 | 7 - 8 | 2474 | 62 | |
| GenBank mRNA | BT149862.1 | - | 7 - 8 | 1570 | 62 | |
More Information
Set Membership: |
|

hth
Genomic Map