Assay Details
| Gene Symbol: MED7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169357.2 | NP_731500.1 | 1 - 2 | 430 | 116 | |
| GenBank mRNA | AF293979.1 | - | 1 - 2 | 377 | 116 | |
| BT023304.1 | - | 1 - 2 | 430 | 116 | ||
More Information
Set Membership: |
|

MED7
Genomic Map