Assay Details
| Gene Symbol: cu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104275.3 | NP_001097745.1 | 5 - 6 | 1323 | 70 | |
| NM_001104276.2 | NP_001097746.1 | 5 - 6 | 1805 | 70 | ||
| NM_001104277.2 | NP_001097747.1 | 6 - 7 | 1478 | 70 | ||
| NM_001275542.1 | NP_001262471.1 | 5 - 6 | 1211 | 70 | ||
| NM_169374.2 | NP_731539.2 | 7 - 8 | 1583 | 70 | ||
| NM_169375.2 | NP_731540.2 | 5 - 6 | 1308 | 70 | ||
| GenBank mRNA | BT023825.1 | - | 6 - 7 | 1471 | 70 | |
| BT025859.1 | - | 6 - 7 | 1448 | 70 | ||
| BT070224.1 | - | 5 - 6 | 1308 | 70 | ||
More Information
Set Membership: |
|

cu
Genomic Map